3-Ethyl-1-propyl-8-(1-(3-(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-3,7-dihydro-1H-purine-2,6-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: GS-6201

A302129-5MG A302129-50MG A302129-100MG A302129-10MG A302129-250MG
77668-340EA 60.55 USD
77668-340 77872-414 77872-411 77872-412 77872-413
3-Ethyl-1-propyl-8-(1-(3-(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-3,7-dihydro-1H-purine-2,6-dione ≥98%
3-Ethyl-1-propyl-8-(1-(3-(trifluoromethyl)benzyl)-1H-pyrazol-4-yl)-3,7-dihydro-1H-purine-2,6-dione

Precaución: for Research Use Only

Formula: C₂₁H₂₁F₃N₆O₂
MDL Number: MFCD11042420
CAS Number: 752222-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR