WAY-600 ≥99%

Supplier: Ambeed
Warning

Synonyms: 4-(6-(1H-Indol-5-yl)-1-(1-(pyridin-3-ylmethyl)piperidin-4-yl)-1H-pyrazolo[3,4-d]pyrimidin-4-yl)morpholine, WAY600, 4-[6-(1H-indol-5-yl)-1-[1-(pyridin-3-ylmethyl)piperidin-4-yl]pyrazolo[3,4-d]pyrimidin-4-yl]morpholine, WAY-600

A301338-10MG A301338-2MG A301338-5MG
77685-858EA 77 USD
77685-858 77685-859 77685-860
WAY-600 ≥99%
WAY-600

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₈O
MW: 494.59 g/mol
MDL Number: MFCD22419019
CAS Number: 1062159-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR