7-Amino-1,2,3,4-tetrahydroquinoline ≥98%

Supplier: Ambeed
Warning

A300427-5G A300427-250MG
77746-564EA 222 USD
77746-564 77746-563
7-Amino-1,2,3,4-tetrahydroquinoline ≥98%
7-Amino-1,2,3,4-tetrahydroquinoline

Precaución: for Research Use Only

Formula: C₉H₁₂N₂
MW: 148.20 g/mol
MDL Number: MFCD08063884
CAS Number: 153856-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR