2'-Bromo-2,2,2-trifluoroacetophenone ≥95%

Supplier: Ambeed
Warning

A300217-100MG A300217-250MG A300217-5G
77883-823EA 50 USD
77883-823 77883-824 77883-825
2'-Bromo-2,2,2-trifluoroacetophenone ≥95%
2'-Bromo-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O
MDL Number: MFCD07380806
CAS Number: 244229-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR