(S)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene ≥98%, ee 99%, Ambeed.Inc

Supplier: Ambeed
Warning

A298605-1G A298605-5G
77593-814EA 44.08 USD
77593-814 77683-890
(S)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene ≥98%, ee 99%, Ambeed.Inc
(S)-3,3'-Diiodo-2,2'-bis(methoxymethoxy)-1,1'-binaphthalene

Precaución: For research use only.

Formula: C₂₄H₂₀I₂O₄
MW: 626.22 g/mol
MDL Number: MFCD17018518
CAS Number: 219583-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR