(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid ≥98%

Supplier: Ambeed
Warning

A298281-100MG A298281-250MG
77879-585EA 85 USD
77879-585 77879-586
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid ≥98%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-(trifluoromethyl)phenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₂₀F₃NO₄
MDL Number: MFCD03428039
CAS Number: 517905-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR