Silicon(IV) phthalocyanine dichloride ≥90%

Supplier: Ambeed
Warning

Synonyms: Silicon phthalocyanine dichloride

A297977-250MG A297977-10G A297977-1G
77612-714EA 36.05 USD
77612-714 77683-886 77683-887
Silicon(IV) phthalocyanine dichloride ≥90%
Silicon(IV) phthalocyanine dichloride

Precaución: For research use only.

Formula: C₃₂H₁₆Cl₂N₈Si
MW: 611.5145 g/mol
MDL Number: MFCD00010723
CAS Number: 19333-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR