Methyl 4-oxochroman-7-carboxylate ≥97%

Supplier: Ambeed
Warning

A297601-100MG A297601-250MG
77883-810EA 30 USD
77883-810 77883-811
Methyl 4-oxochroman-7-carboxylate ≥97%
Methyl 4-oxochroman-7-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀O₄
MDL Number: MFCD14706083
CAS Number: 41118-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR