Phthaloyl-Leu-OH ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(1,3-Dioxoisoindolin-2-yl)-4-methylpentanoic acid, Pht-Leu-OH

A297247-100G A297247-5G A297247-500G A297247-25G
77685-848EA 351 USD
77685-848 77685-849 77902-063 77900-974
Phthaloyl-Leu-OH ≥98%
Phthaloyl-Leu-OH

Precaución: for Research Use Only

Formula: C₁₄H₁₅NO₄
MDL Number: MFCD00065026
CAS Number: 2419-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR