2,5-Dichloro-3-(5-(3,4-dihydroxy-5-nitrophenyl)-1,2,4-oxadiazol-3-yl)-4,6-dimethylpyridine 1-oxide 98+%

Supplier: Ambeed
Warning

A297021-10MG A297021-1MG A297021-5MG
77616-290EA 88.52 USD
77616-290 77616-292 77616-294
2,5-Dichloro-3-(5-(3,4-dihydroxy-5-nitrophenyl)-1,2,4-oxadiazol-3-yl)-4,6-dimethylpyridine 1-oxide 98+%
2,5-Dichloro-3-(5-(3,4-dihydroxy-5-nitrophenyl)-1,2,4-oxadiazol-3-yl)-4,6-dimethylpyridine 1-oxide

Precaución: for Research Use Only

Formula: C₁₅H₁₀Cl₂N₄O₆
MDL Number: MFCD19443745
CAS Number: 923287-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR