tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate ≥95%

Supplier: Ambeed
Danger

Synonyms: 1-Boc-3-(2-aminoethyl)piperidine

A296720-100MG A296720-250MG
77881-815EA 43 USD
77881-815 77881-816
tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate ≥95%
tert-Butyl 3-(2-aminoethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD04038458
CAS Number: 259180-77-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR