2',4'-Dichloroacetophenone ≥98%

Supplier: Ambeed
Warning

Synonyms: 2′,4′-Dichloroacetophenone

A296330-100G A296330-500G A296330-1000G
77616-286EA 23.69 USD
77616-286 77616-288 77746-553
2',4'-Dichloroacetophenone ≥98%
2',4'-Dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
MDL Number: MFCD00000581
CAS Number: 2234-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR