6-Chloro-5-methoxyindoline-2,3-dione ≥97%

Supplier: Ambeed
Warning

A295995-100MG A295995-250MG
77685-841EA 76 USD
77685-841 77685-842
6-Chloro-5-methoxyindoline-2,3-dione ≥97%
6-Chloro-5-methoxyindoline-2,3-dione

Precaución: for Research Use Only

Formula: C₉H₆ClNO₃
MDL Number: MFCD04114068
CAS Number: 85778-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR