3-(tert-Butyl)-4-hydroxybenzonitrile ≥97%
Supplier: Ambeed
|
Warning
|
A295533-1G
A295533-5G
77612-706EA
105.06
USD
77612-706
77885-785
3-(tert-Butyl)-4-hydroxybenzonitrile ≥97%
3-(tert-Butyl)-4-hydroxybenzonitrile
Precaución: for Research Use Only
Formula:
C₁₁H₁₃NO |
MDL Number:
MFCD16999179 CAS Number: 4910-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...