(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-cyclohexylpropanoate ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-Cha-OMe

A295454-1G A295454-5G
77601-318EA 23.58 USD
77601-318 77601-320
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-cyclohexylpropanoate ≥95%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-cyclohexylpropanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₄
MW: 285.37 g/mol
MDL Number: MFCD00191183
CAS Number: 98105-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR