N-((1R,2R)-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-1-hydroxy-3-(pyrrolidin-1-yl)propan-2-yl)octanamide ((2R,3R)-2,3-dihydroxysuccinate)(2:1) ≥98%

Supplier: Ambeed
Warning

Synonyms: Eliglustat hemitartrate

A295388-5MG A295388-10MG A295388-1MG
77616-282EA 30.9 USD
77616-282 77616-278 77616-280
N-((1R,2R)-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-1-hydroxy-3-(pyrrolidin-1-yl)propan-2-yl)octanamide ((2R,3R)-2,3-dihydroxysuccinate)(2:1) ≥98%
N-((1R,2R)-1-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)-1-hydroxy-3-(pyrrolidin-1-yl)propan-2-yl)octanamide ((2R,3R)-2,3-dihydroxysuccinate)(2:1)

Precaución: for Research Use Only

Formula: C₅₀H₇₈N₄O₁₄
MW: 959.17 g/mol
MDL Number: MFCD22665712
CAS Number: 928659-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR