2-(Bromomethyl)acrylic acid ≥98%

Supplier: Ambeed
Danger

A295346-1G A295346-250MG A295346-5G
77883-800EA 88 USD
77883-800 77883-801 77883-802
2-(Bromomethyl)acrylic acid ≥98%
2-(Bromomethyl)acrylic acid

Precaución: for Research Use Only

Formula: C₄H₅BrO₂
MDL Number: MFCD00004259
CAS Number: 72707-66-5
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR