Copper(I) trifluoromethanethiolate ≥95%

Supplier: Ambeed
Warning

Synonyms: (Trifluoromethylthio)copper(I), Copper(I) Trifluoromethanethiolate, Trifluoromethanethiol Copper(I) Salt

A295142-250MG A295142-5G
77626-514EA 92.7 USD
77626-514 77879-580
Copper(I) trifluoromethanethiolate ≥95%
Copper(I) trifluoromethanethiolate

Precaución: for Research Use Only

Formula: CCuF₃S
MDL Number: MFCD00059046
CAS Number: 3872-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR