rel-(6R,7S,Z)-3-Butylidene-6,7-dihydroxy-4,5,6,7-tetrahydroisobenzofuran-1(3H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Senkyunolide H

A295128-1MG A295128-5MG A295128-10MG
77612-702EA 55.62 USD
77612-702 77612-704 77612-700
rel-(6R,7S,Z)-3-Butylidene-6,7-dihydroxy-4,5,6,7-tetrahydroisobenzofuran-1(3H)-one ≥98%
rel-(6R,7S,Z)-3-Butylidene-6,7-dihydroxy-4,5,6,7-tetrahydroisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.25 g/mol
MDL Number: MFCD32182625
CAS Number: 94596-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR