(2-Ethynylphenyl)methanol ≥95%

Supplier: Ambeed
Danger

A293918-1G A293918-250MG A293918-5G
77598-402EA 59.45 USD
77598-402 77616-274 77748-574
(2-Ethynylphenyl)methanol ≥95%
(2-Ethynylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD06200681
CAS Number: 10602-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR