Oxazolidine-2,5-dione ≥97%

Supplier: Ambeed
Warning

A293784-100MG A293784-5G
77746-548EA 15 USD
77746-548 77746-549
Oxazolidine-2,5-dione ≥97%
Oxazolidine-2,5-dione

Precaución: for Research Use Only

Formula: C₃H₃NO₃
MW: 101.06 g/mol
MDL Number: MFCD00075543
CAS Number: 2185-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR