tert-Butyl 3-(((benzyloxy)carbonyl)amino)propanoate ≥95%

Supplier: Ambeed
Warning

A293564-1G A293564-25G A293564-5G
77885-775EA 41 USD
77885-775 77885-776 77885-777
tert-Butyl 3-(((benzyloxy)carbonyl)amino)propanoate ≥95%
tert-Butyl 3-(((benzyloxy)carbonyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD03837779
CAS Number: 18605-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR