N,N-Diethyl-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥96%

Supplier: Ambeed
Warning

A293509-1G A293509-250MG A293509-5G
77883-791EA 189 USD
77883-791 77883-792 77883-793
N,N-Diethyl-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide ≥96%
N,N-Diethyl-2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₇H₂₅BFNO₃
MDL Number: MFCD11855963
CAS Number: 1150271-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR