(6R,7R)-4-Methoxybenzyl-3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥97%

Supplier: Ambeed
Danger

Synonyms: 4-Methoxybenzyl 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylate, 3-Chloromethyl-7-(2-phenylacetamido)-3-cephem-4-carboxylic acid 4-Methoxybenzyl ester, GCLE, (6R,7R)-4-Methoxybenzyl 3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A292248-100G A292248-25G
77593-788EA 23.58 USD
77593-788 77601-298
(6R,7R)-4-Methoxybenzyl-3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate ≥97%
(6R,7R)-4-Methoxybenzyl-3-(chloromethyl)-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₄H₂₃ClN₂O₅S
MW: 486.967 g/mol
MDL Number: MFCD00191253
CAS Number: 104146-10-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR