(S)-N-((S)-1-Cyano-2-(4-(3-methyl-2-oxo-2,3-dihydrobenzo[d]oxazol-5-yl)phenyl)ethyl)-1,4-oxazepane-2-carboxamide ≥98%

Supplier: Ambeed
Danger

Synonyms: Brensocatib

A291759-50MG A291759-100MG A291759-10MG
77616-268EA 309 USD
77616-268 77748-567 77748-568
(S)-N-((S)-1-Cyano-2-(4-(3-methyl-2-oxo-2,3-dihydrobenzo[d]oxazol-5-yl)phenyl)ethyl)-1,4-oxazepane-2-carboxamide ≥98%
(S)-N-((S)-1-Cyano-2-(4-(3-methyl-2-oxo-2,3-dihydrobenzo[d]oxazol-5-yl)phenyl)ethyl)-1,4-oxazepane-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₄N₄O₄
MW: 420.46 g/mol
MDL Number: MFCD31619232
CAS Number: 1802148-05-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR