5-Bromo-2-fluorophenylhydrazine hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
A291536-10G
A291536-1G
A291536-5G
77892-584EA
348
USD
77892-584
77892-585
77892-586
5-Bromo-2-fluorophenylhydrazine hydrochloride ≥97%
5-Bromo-2-fluorophenylhydrazine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₆BrFN₂·ClH |
MDL Number:
MFCD03094411 CAS Number: 214916-08-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...