1-(2-(Azocan-1-yl)ethyl)guanidine sulfate ≥98%

Supplier: Ambeed
Warning

Synonyms: Guanethidine sulphate

A291526-100MG A291526-1MG
77601-296EA 29.73 USD
77601-296 77668-292
1-(2-(Azocan-1-yl)ethyl)guanidine sulfate ≥98%
1-(2-(Azocan-1-yl)ethyl)guanidine sulfate

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₄O₄S
MW: 296.39 g/mol
MDL Number: MFCD00035403
CAS Number: 645-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR