(5-Phenylisoxazol-3-yl)methanamine hydrochloride ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A291448-5G A291448-1G
77683-876EA 1049 USD
77683-876 77890-539
(5-Phenylisoxazol-3-yl)methanamine hydrochloride ≥98%, Ambeed.Inc
(5-Phenylisoxazol-3-yl)methanamine hydrochloride

Precaución: For research use only.

Formula: C₁₀H₁₁ClN₂O
MDL Number: MFCD08062591
CAS Number: 1187928-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR