Fmoc-Pen(Acm)-OH ≥98%

Supplier: Ambeed
Warning

A291020-100MG A291020-1G A291020-250MG
77879-569EA 46 USD
77879-569 77879-570 77879-571
Fmoc-Pen(Acm)-OH ≥98%
Fmoc-Pen(Acm)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₅S
MW: 442.53 g/mol
MDL Number: MFCD00151935
CAS Number: 201531-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR