7-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥98%

Supplier: Ambeed
Warning

A290986-10G A290986-25G A290986-5G
77892-580EA 101 USD
77892-580 77892-581 77892-582
7-Nitro-2H-1,4-benzoxazin-3(4H)-one ≥98%
7-Nitro-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O₄
MDL Number: MFCD00090355
CAS Number: 81721-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR