2-Methoxy-3-(1-methyl-1H-1,2,4-triazol-3-yl)aniline ≥97%

Supplier: Ambeed
Warning

A290160-5G A290160-250MG
77571-792EA 91.67 USD
77571-792 77746-538
2-Methoxy-3-(1-methyl-1H-1,2,4-triazol-3-yl)aniline ≥97%
2-Methoxy-3-(1-methyl-1H-1,2,4-triazol-3-yl)aniline

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₄O
MW: 204.23 g/mol
MDL Number: MFCD30489435
CAS Number: 1609394-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR