Furo[2,3-c]pyridine-7(6H)-one ≥95%

Supplier: Ambeed
Warning

A289913-100MG A289913-250MG
77883-782EA 121 USD
77883-782 77883-783
Furo[2,3-c]pyridine-7(6H)-one ≥95%
Furo[2,3-c]pyridine-7(6H)-one

Precaución: for Research Use Only

Formula: C₇H₅NO₂
MDL Number: MFCD01544382
CAS Number: 84400-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR