1-(4-Chloro-2-fluoro-6-methoxyphenyl)ethanone ≥95%
Supplier: Ambeed
|
Warning
|
A289330-1G
A289330-5G
77683-865EA
169
USD
77683-865
77683-866
1-(4-Chloro-2-fluoro-6-methoxyphenyl)ethanone ≥95%
1-(4-Chloro-2-fluoro-6-methoxyphenyl)ethanone
Precaución: For research use only.
Formula:
C₉H₈ClFO₂ MW: 202.61 g/mol |
CAS Number:
1822668-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...