5-Fluoropyridine-2-sulfonyl chloride ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

A289299-1G A289299-250MG A289299-100MG
77683-864EA 1481 USD
77683-864 77892-576 77892-575
5-Fluoropyridine-2-sulfonyl chloride ≥95%, Ambeed.Inc
5-Fluoropyridine-2-sulfonyl chloride

Precaución: For research use only.

Formula: C₅H₃ClFNO₂S
MDL Number: MFCD13189069
CAS Number: 1060802-47-2
UN: 2922
ADR: 8, 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR