(S)-2-Amino-N-((R)-3-methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butyl)-3-phenylpropanamide hydrochloride ≥97%

Supplier: Ambeed
Warning

A288460-100MG A288460-1G A288460-250MG A288460-5G
77881-795EA 53 USD
77881-795 77881-796 77881-797 77881-798
(S)-2-Amino-N-((R)-3-methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butyl)-3-phenylpropanamide hydrochloride ≥97%
(S)-2-Amino-N-((R)-3-methyl-1-((3aS,4S,6S,7aR)-3a,5,5-trimethylhexahydro-4,6-methanobenzo[d][1,3,2]dioxaborol-2-yl)butyl)-3-phenylpropanamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₃₈BClN₂O₃
MW: 448.83 g/mol
MDL Number: MFCD18252917
CAS Number: 205393-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR