2-Methyl-2H-indazole-5-boronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Methyl-2H-indazol-5-yl)boronic acid

A288191-5G A288191-1G
77571-786EA 187 USD
77571-786 77748-559
2-Methyl-2H-indazole-5-boronic acid ≥98%
2-Methyl-2H-indazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD09870056
CAS Number: 952319-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR