5-Bromogramine ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(5-Bromo-1H-indol-3-yl)-N,N-dimethylmethanamine

A288062-5G A288062-1G A288062-25G
77598-398EA 149.65 USD
77598-398 77626-488 77626-490
5-Bromogramine ≥98%
5-Bromogramine

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂
MW: 253.14 g/mol
MDL Number: MFCD00055980
CAS Number: 830-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR