(S)-6-(2-(Benzo[d][1,3]dioxol-5-yl)ethyl)-4-methoxy-5,6-dihydro-2H-pyran-2-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Dihydromethysticin

A288033-1MG A288033-10MG A288033-5MG
77601-282EA 76.88 USD
77601-282 77668-276 77668-278
(S)-6-(2-(Benzo[d][1,3]dioxol-5-yl)ethyl)-4-methoxy-5,6-dihydro-2H-pyran-2-one ≥98%
(S)-6-(2-(Benzo[d][1,3]dioxol-5-yl)ethyl)-4-methoxy-5,6-dihydro-2H-pyran-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₅
MW: 276.28 g/mol
MDL Number: MFCD00221731
CAS Number: 19902-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR