Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II) 99.99% (metals basis)

Supplier: Ambeed
Danger

A288004-1G A288004-5G A288004-250MG
77626-484EA 46.35 USD
77626-484 77626-486 77877-947
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II) 99.99% (metals basis)
Bis(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionate)copper(II)

Precaución: for Research Use Only

Formula: Cu(C₁₀H₁₀F₇O₂)₂
MDL Number: MFCD00156516
CAS Number: 80289-21-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR