1-((3aR,4S,9bS)-rel-4-(6-Bromobenzo[d][1,3]dioxol-5-yl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-8-yl)ethanone ≥98%

Supplier: Ambeed
Danger

Synonyms: G-1

A287698-10MG A287698-1MG A287698-5MG
77616-232EA 70.04 USD
77616-232 77616-234 77616-236
1-((3aR,4S,9bS)-rel-4-(6-Bromobenzo[d][1,3]dioxol-5-yl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-8-yl)ethanone ≥98%
1-((3aR,4S,9bS)-rel-4-(6-Bromobenzo[d][1,3]dioxol-5-yl)-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-8-yl)ethanone

Precaución: for Research Use Only

Formula: C₂₁H₁₈BrNO₃
MW: 412.27 g/mol
MDL Number: MFCD16618392
CAS Number: 881639-98-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR