N-Boc-N-methyl-3-chloro-1-propanamine ≥95%

Supplier: Ambeed
Warning

A287684-1G A287684-250MG
77746-529EA 140 USD
77746-529 77746-530
N-Boc-N-methyl-3-chloro-1-propanamine ≥95%
N-Boc-N-methyl-3-chloro-1-propanamine

Precaución: for Research Use Only

Formula: C₉H₁₈ClNO₂
MW: 207.70 g/mol
MDL Number: MFCD09031544
CAS Number: 114326-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR