3-Chloro-5-methyl-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-3-methyl-1H-pyrazole

A287618-1G A287618-250MG A287618-5G
77877-943EA 35 USD
77877-943 77877-944 77877-945
3-Chloro-5-methyl-1H-pyrazole ≥98%
3-Chloro-5-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂
MW: 116.55 g/mol
MDL Number: MFCD00233972
CAS Number: 15953-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR