tert-Butyl 3-(2-(2-(methylamino)ethoxy)ethoxy)propanoate ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methylamino-PEG2-Boc
A287551-100MG
A287551-250MG
77616-230EA
39.14
USD
77616-230
77748-557
tert-Butyl 3-(2-(2-(methylamino)ethoxy)ethoxy)propanoate ≥97%
tert-Butyl 3-(2-(2-(methylamino)ethoxy)ethoxy)propanoate
Precaución: for Research Use Only
Formula:
C₁₂H₂₅NO₄ MW: 247.33 g/mol |
MDL Number:
MFCD28155207 CAS Number: 1807521-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...