tert-Butyl (cis-3-aminocyclopentyl)carbamate ≥98%

Supplier: Ambeed
Warning

A287375-100MG A287375-1G A287375-250MG
77885-758EA 56 USD
77885-758 77885-759 77885-760
tert-Butyl (cis-3-aminocyclopentyl)carbamate ≥98%
tert-Butyl (cis-3-aminocyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD18837747
CAS Number: 454709-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR