Phenethyl butyrate ≥99%

Supplier: Ambeed
Warning

A287292-25G A287292-100G A287292-500G
77601-270EA 13.33 USD
77601-270 77879-557 77879-558
Phenethyl butyrate ≥99%
Phenethyl butyrate

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD00048718
CAS Number: 103-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR