2-Chloro-4-fluoro-5-methoxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-4-fluoro-5-methoxyphenyl)boronic acid

A287247-1G A287247-250MG A287247-100MG
77612-652EA 183.34 USD
77612-652 77612-654 77683-862
2-Chloro-4-fluoro-5-methoxyphenylboronic acid ≥98%
2-Chloro-4-fluoro-5-methoxyphenylboronic acid

Precaución: For research use only.

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
MDL Number: MFCD09998845
CAS Number: 1256355-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR