2-Oxa-5-azaspiro[3.4]octane hemioxalate ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A286739-250MG A286739-100MG A286739-1G A286739-50MG
77683-861EA 352 USD
77683-861 77890-526 77890-527 77890-528
2-Oxa-5-azaspiro[3.4]octane hemioxalate ≥97%, Ambeed.Inc
2-Oxa-5-azaspiro[3.4]octane hemioxalate

Precaución: For research use only.

Formula: C₁₄H₂₄N₂O₆
MDL Number: MFCD22373635
CAS Number: 1380571-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR