Chlorhexidine gluconate 20% (w/w) in water

Supplier: Ambeed
Danger

A286403-100G A286403-500G
77593-754EA 15.38 USD
77593-754 77601-260
Chlorhexidine gluconate 20% (w/w) in water
Chlorhexidine gluconate

Precaución: for Research Use Only

Formula: C₂₂H₃₀Cl₂N₁₀·2C₆H₁₂O₇
MDL Number: MFCD00083599
CAS Number: 18472-51-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR