Diindium trisulphate 99.99%

Supplier: Ambeed
Warning

Synonyms: Diindium tris(sulphate), Indium(III) sulphate, Indium (III) sulphate

A285449-25G A285449-5G A285449-1G A285449-100G
77601-254EA 41 USD
77601-254 77601-256 77668-266 77879-555
Diindium trisulphate 99.99%
Diindium trisulphate

Precaución: for Research Use Only

Formula: O₁₂In₂S₃
MDL Number: MFCD00011059
CAS Number: 13464-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR