Beta-elemonicacid ≥95%

Supplier: Ambeed
Warning

Synonyms: β-Elemonic acid

A284576-1MG A284576-25MG A284576-5MG
77593-746EA 10.25 USD
77593-746 77593-748 77593-750
Beta-elemonicacid ≥95%
Beta-elemonicacid

Precaución: for Research Use Only

Formula: C₃₀H₄₆O₃
MW: 454.68 g/mol
MDL Number: MFCD00467083
CAS Number: 28282-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR